missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD54 / ICAM-1 Mouse, Clone: W-CAM-1; same as Wehi-CAM-1 or 1H4, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3383-MSM1-P1
This item is not returnable.
View return policy
Specifications
CD54 / ICAM-1 | |
Cell Surface | |
W-CAM-1; same as Wehi-CAM-1 or 1H4 | |
Unconjugated | |
NP_000192 | |
Mouse | |
85-115kDa | |
Protein A/G Purified | |
RUO | |
3383 | |
IgG2b Îş |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Monoclonal | |
200ÎĽg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide | |
ICAM1 | |
Raji Burkitt lymphoma cells | |
Raji cells, MOLT 4, HeLa or SW480 cells. Human tonsil. | |
100 ÎĽg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD54 / ICAM-1 Mouse, Clone: W-CAM-1; same as Wehi-CAM-1 or 1H4, enQuireBio™ > 100 μg
Spot an opportunity for improvement?Share a Content Correction