missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Mouse anti-Human, BUV395, Clone: MY31, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742663
6120.40 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow Cytometry.
Specifications
CD56 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
MY31 | |
Brilliant Ultraviolet 395 | |
50 ÎĽg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD56 Mouse anti-Human, BUV395, Clone: MY31, BD OptiBuild™ > Hu CD56 BUV395 MY31 50 μg
Spot an opportunity for improvement?Share a Content Correction