Läs mer
CD71 (Transferrin Receptor) Mouse, anti-Human, APC, Clone: OKT9, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60106AZ
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- The OKT9 antibody reacts with CD71 (transferrin receptor), an ∼180 - 190 kDa disulfide-bonded homodimer and type 2 transmembrane glycoprotein expressed at high levels on a broad range of actively proliferating cells, as well as reticulocytes, monocytes, macrophages and marrow stromal cells
- Surface levels are low on resting leukocytes but are upregulated on lymphocytes, monocytes and macrophages following antigen or mitogen stimulation
- Expression is lost during differentiation of reticulocytes into mature erythrocytes
- CD71 mediates uptake of transferrin–iron complexes through receptor-mediated endocytosis and recycling of the apotransferrin–receptor complex to the cell surface
- It has also served as a target for drug delivery
- The OKT9 antibody binds an epitope that reportedly overlaps with that of antibody clone CY1G4 but is distinct from the ligand binding site for transferrin
Specifikationer
| CD71 (Transferrin Receptor) | |
| Monoclonal | |
| APC | |
| 100 Tests | |
| Human | |
| Erythroid Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OKT9 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.