missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD79b (B-Cell Marker) Mouse, Clone: IGB/1844, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 974-MSM4-P0
This item is not returnable.
View return policy
Specifications
CD79b (B-Cell Marker) | |
Cell surface | |
IGB/1844 | |
Unconjugated | |
Mouse | |
39kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
974 | |
IgG1 κ |
Immunohistochemistry, ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human CD79b protein fragment (apprx. aa29-159) | |
Daudi or Ramos cells. Germinal center B-cells in a lymph node or tonsil. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD79b (B-Cell Marker) Mouse, Clone: IGB/1844, enQuireBio™ > 20 μg
Spot an opportunity for improvement?Share a Content Correction