missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BUV563, Clone: RPA-T8, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612915
Additional Details : Weight : 0.09000kg
Description
CD8 Monoclonal antibody specifically detects CD8 in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifications
CD8 | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
25 Tests | |
Primary | |
Human, Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
RPA-T8 | |
Mouse | |
RUO | |
CD8 | |
Refrigerate (2 to 8°C) |