missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse anti-Human , PE-CF594, Clone: HIT8a, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566860
3845.58 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry
Specifications
CD8 | |
Monoclonal | |
PE-CF594 | |
CD8α; CD8A; CD8 alpha; Leu2a; MAL; T8; p32 | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |
Flow Cytometry | |
HIT8a | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG1 Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD8 Mouse anti-Human , PE-CF594, Clone: HIT8a, BD Biosciences™ > 100 Tests; PE-CF594
Spot an opportunity for improvement?Share a Content Correction