missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD80 (B7-1) Mouse anti-Human, BV605, Clone: 2D10.4 (also known as 2D10), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751721
6019.82 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD80 (B7-1) Monoclonal antibody specifically detects CD80 (B7-1) in Human samples. It is validated for Flow Cytometry.

Specifications
CD80 (B7-1) | |
Monoclonal | |
0.2 mg/mL | |
Hu CD80 (B7-1) BV605 2D10.4 50 μg | |
Not Reported | |
RUO | |
CD80 (B7-1) | |
Refrigerate (2 to 8°C) | |
Human |
Flow Cytometry | |
2D10.4 (also known as 2D10) | |
Brilliant Violet 605 | |
Mouse | |
1 x 50 μg | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD80 (B7-1) Mouse anti-Human, BV605, Clone: 2D10.4 (also known as 2D10), BD OptiBuild™ > Quantity: 1 x 50 μg
Spot an opportunity for improvement?Share a Content Correction