Learn More
CD97 Mouse anti-Human, Unconjugated, Clone: 3F10C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66972-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889305
  • 1656.73 SEK / 20µL
Estimated Shipment: 26-04-2024
to see stock.
Description
Description
The CD97 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD97. This antibody recognizes human, mouse antigen. The CD97 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
CD97
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, Flow Cytometry, ELISA
3F10C2
Unconjugated
20 μL
976
IgG2a