Learn More
CD97 Mouse anti-Human, Unconjugated, Clone: 3F10C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66972-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The CD97 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD97. This antibody recognizes human, mouse antigen. The CD97 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
CD97 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Flow Cytometry, ELISA | |
3F10C2 | |
Unconjugated | |
150 μL | |
976 | |
IgG2a |