missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Mouse anti-Human, Clone: 5B6A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60354-1-IG-150UL
This item is not returnable.
View return policy
Description
CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell adhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the hematopoietic system to T, NK and myeloid cells.Specifications
CD99 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14209 | |
CD99 | |
CD99 Fusion Protein Ag19474 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
5B6A4 | |
Unconjugated | |
CD99 | |
12E7, CD99, CD99 antigen, CD99 molecule, E2 antigen, MIC2, MIC2X, MIC2Y, Protein MIC2, T cell surface glycoprotein E2 | |
Mouse | |
Protein G | |
RUO | |
4267 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD99 Mouse anti-Human, Clone: 5B6A4, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction