Learn More
CDC14B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25187-1-AP-20UL
 View more versions of this product
Product Code. 16883745
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
CDC14B is a member of the dual specificity protein tyrosine phosphatase family. This protein is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, which suggests the role in cell cycle control. This protein has been shown to interact with and dephosphorylates tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splice of this gene results in 3 transcript variants encoding distinct isoforms.
Specifications
Specifications
CDC14B
Polyclonal
Unconjugated
CDC14B
CDC14B, Cdc14B1, Cdc14B2, CDC14B3, hCDC14B
Rabbit
Antigen Affinity Chromatography
RUO
8555
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60729
CDC14B
CDC14B Fusion Protein Ag18227
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDC14B Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated