Learn More
CDC27 APC3 Mouse anti-Human, Unconjugated, Clone: 2F10G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67239-1-IG-20UL
 View more versions of this product
Product Code. 16889755
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The CDC27; APC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC27; APC3. This antibody recognizes human, mouse, rat antigen. The CDC27; APC3 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CDC27 APC3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
2F10G2
Unconjugated
20 μL
996
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDC27 APC3 Mouse anti-Human, Unconjugated, Clone: 2F10G2, Proteintech > 20 μL