Learn More
CDC27 APC3 Mouse anti-Human, Unconjugated, Clone: 2F10G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67239-1-IG-150UL
 View more versions of this product
Product Code. 16879755
  • 4652.90 SEK / 150µL
Description
The CDC27; APC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC27; APC3. This antibody recognizes human, mouse, rat antigen. The CDC27; APC3 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CDC27 APC3
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
2F10G2
Unconjugated
150 ÎĽL
996
IgG2b
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CDC27 APC3 Mouse anti-Human, Unconjugated, Clone: 2F10G2, Proteintech > 150 ÎĽL