Learn More
CDCA8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12465-1-AP-150UL
 View more versions of this product
Product Code. 16880184
  • 4652.90 SEK / 150µL
Description
CDCA8 is a component of a chromosomal passenger complex (CPC) required for stability of the bipolar mitotic spindle. The chromosomal passenger complex, which includes Survivin, CDCA8, INCENP and Aurora-B, is known to play crucial roles during mitosis and cell division. It was found that CDCA8 interacting with Aurora-B, INCENP and Survivin, increases during G2/M phase and then reduces after exit from M phase. CDCA8 is cell cycle regulated, down-regulated in response to p53/Rb-signaling, and up-regulated in many types of cancerous tissues. In Drosophila cells, inactivation of CDCA8 results in polyploidy, delayed mitosis and abnormal tissue development, indicating its critical role for cell proliferation. Recent studies show that CDCA8 is essential for cell proliferation during early embryonic development, and its early embryonic lethality cannot be rescued by the loss of p53. Its aberrant expression is linked to a poor prognosis for gastric cancer.
Specifications
Specifications
CDCA8
Polyclonal
Unconjugated
CDCA8
BOR, BOREALIN, CDCA8, Dasra B, hDasra B, PESCRG3
Rabbit
Antigen Affinity Chromatography
RUO
500545, 52276, 55143
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q53HL2, Q6AXW0, Q8BHX3
Cdca8
CDCA8 Fusion Protein Ag3136
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
CDCA8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated