missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK2AP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15423-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene.Specifications
CDK2AP2 | |
Polyclonal | |
Unconjugated | |
CDK2AP2 | |
CDK2 associated protein 2, CDK2AP2, DOC 1 related protein, DOC 1R, DOC1R, p14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10263 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75956 | |
CDK2AP2 | |
CDK2AP2 Fusion Protein Ag7730 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |