Learn More
CDK4 Mouse anti-Human, Unconjugated, Clone: 1G2C12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66950-1-IG-150UL
 View more versions of this product
Product Code. 16879265
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The CDK4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK4. This antibody recognizes human, mouse, rat antigen. The CDK4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CDK4
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry
1G2C12
Unconjugated
150 μL
1019
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDK4 Mouse anti-Human, Unconjugated, Clone: 1G2C12, Proteintech > 150 μL