missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21655-1-AP-150UL
This item is not returnable.
View return policy
Description
Could play a role in the terminal differentiation of the intestine.Specifications
CDX1 | |
Polyclonal | |
Unconjugated | |
CDX1 | |
caudal type homeobox 1, Caudal type homeobox protein 1, CDX1, Homeobox protein CDX 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1044, 12590 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18111, P47902 | |
CDX1 | |
CDX1 Fusion Protein Ag16376 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction