Learn More
CEBPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12997-1-AP-20UL
 View more versions of this product
Product Code. 16880994
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements.
Specifications
Specifications
CEBPG
Polyclonal
Unconjugated
CEBPG
C/EBP gamma, CEBPG, GPE1BP, IG/EBP 1
Rabbit
Antigen Affinity Chromatography
RUO
1054, 12611, 25301
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P26801, P53567, P53568
CEBPG
CEBPG Fusion Protein Ag3663
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CEBPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated