Learn More
CEBPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12997-1-AP-20UL
 View more versions of this product
Product Code. 16880994
  • 1645.54 SEK / 20µL
Description
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements.
Specifications
Specifications
CEBPG
Polyclonal
Unconjugated
CEBPG
C/EBP gamma, CEBPG, GPE1BP, IG/EBP 1
Rabbit
Antigen Affinity Chromatography
RUO
1054, 12611, 25301
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P26801, P53567, P53568
CEBPG
CEBPG Fusion Protein Ag3663
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CEBPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated