Learn More
Invitrogen™ CellBlox™ Blocking Buffer, 100 Tests
Formulated to block nonspecific binding of Invitrogen™ NovaFluor™ labels with cells, which can result in higher background labeling. Also recommended for cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling.
Brand:  Invitrogen™ B001T03F01
Product Code. 17263675
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Formulated to block nonspecific binding of Invitrogen™ NovaFluor™ labels with cells that can result in higher background labeling
  • CellBlox™ Blocking Buffer is a non-antibody, non-protein–based blocking solution, and should be used every time a NovaFluor dye is used for labeling any cell type to minimize background labeling
  • Recommended for use with cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling
  • Use of CellBlox Blocking Buffer requires minimal change to most flow cytometry staining protocols
  • Add 5 ÎĽL of buffer directly to a cell suspension containing 103 to 108 cells prior to the addition of an antibody, with 100 ÎĽL as a final staining volume
  • Buffer may be added to an antibody cocktail mixture prior to labeling cells, by adding 5 ÎĽL of buffer for every stained sample to be labeled with the antibody cocktail mixture, with 100 ÎĽL as a final staining volume
  • Always use CellBlox Blocking Buffer with NovaFluor dyes when labeling cells for best background reduction
  • Compatible with all fluorophores and with Invitrogen™ LIVE/DEAD™ Fixable Dead Cell Stains
  • Can be used with any fluorophore-antibody conjugate as a high-performance monocyte and macrophage blocking solution
  • CellBlox Blocking Buffer is compatible with other blocking reagents such as Fc Block, blocking proteins, Brilliant Stain Buffer, and Super Bright Complete Staining Buffer
  • Buffer is not required when labeling antibody-capture beads
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3048998 Certificate TypeCertificate of Analysis Date20/09/2024 Catalog NumberB001T03F01
Lot Number2789924 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog NumberB001T03F01
Lot Number2585870 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog NumberB001T03F01
Lot Number2513929 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog NumberB001T03F01
Lot NumberM131516 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog NumberB001T03F01
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ CellBlox™ Blocking Buffer, 100 Tests > 100 tests