Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm are recommended for colony counting, particle testing, and microscopy.
2275.74 SEK
Specifications
Color | Gray/White Grid |
---|---|
Material | Cellulose Nitrate |
Pore Size | 0.45 ÎĽm |
Sterility | Sterile |
Thickness (Metric) | 115 to 145 ÎĽm |
Description
- Sterile, individually packaged filters
- Save preparatory time
- Conform with GLP - filter identification via lot number printed on each envelope.
- Positioned correctly with no slippage under the edge seal
- Perfect grid printing
- Particle-free
- In accordance with ISO 7704, ISO 7899-2, ISO 8199, ISO 9308-1 and EN 12780
- Manufactured for use with Sartorius Nutrient Pad Sets in accordance with many international regulations and recommendations: international pharmacopoeias, DIN and ISO Standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA
- All membranes are made of cellulose nitrate (cellulose ester), a material which assures effective retention with high flow rates and optimum colony growth
- Printed grid divides the filtration area into 130 squares, each measuring 3.1x3.1mm , which makes counting easier, especially for higher bacteria counts and for micro-colonies, but does not influence the growth
- Filter colors allow the best contrast to the colonies or particles
- Outstanding microorganism recovery rate
- High flow membranes
- Sterilized by gamma irradiation
- Compatible with aqueous solutions (pH 4-8), hydrocarbons, and several other organic solvents
Specifications
Gray/White Grid | |
0.45 ÎĽm | |
115 to 145 ÎĽm |
Cellulose Nitrate | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm