missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17007-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
CENPL | |
Polyclonal | |
Unconjugated | |
CENPL | |
C1orf155, CENP L, CENPL, centromere protein L, ICEN33, RP3 383J4.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289150, 70454, 91687 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3U3S3, Q3ZAU7, Q8N0S6 | |
Cenpl | |
CENPL Fusion Protein Ag10703 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |