missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CENPL |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836824
|
Proteintech
17007-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826824
|
Proteintech
17007-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CENPL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q3U3S3, Q3ZAU7, Q8N0S6 | |
289150, 70454, 91687 | |
CENPL Fusion Protein Ag10703 | |
Primary | |
-20°C | |
CENPL |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C1orf155, CENP L, CENPL, centromere protein L, ICEN33, RP3 383J4.1 | |
Cenpl | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |