missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP110 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CEP110 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883805
|
Proteintech
25235-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873805
|
Proteintech
25235-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CEP110 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q7Z7A1 | |
11064 | |
CEP110 Fusion Protein Ag18907 | |
Primary | |
-20°C | |
CNTRL |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
110 kDa centrosomal protein, Centriolin, Centrosomal protein 1, centrosomal protein 110kDa, CEP1, CEP110, FAN | |
CNTRL | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CEP110 Rabbit anti-Human, Polyclonal, Proteintech