missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP192 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1395.65 SEK - 3665.00 SEK
Specifications
Antigen | CEP192 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CEP192 is required for mitotic centrosome and spindle assembly. Appears to be a major regulator of pericentriolar material (PCM) recruitment, centrosome maturation, and centriole duplication.Specifications
CEP192 | |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q8TEP8 | |
55125 | |
Peptide | |
Primary | |
-20°C | |
CEP192 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
centrosomal protein 192kDa, CEP192, KIAA1569, KIAA1569 protein | |
CEP192 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CEP192 Rabbit anti-Human, Polyclonal, Proteintech