missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP97 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CEP97 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881595
|
Proteintech
22050-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16871595
|
Proteintech
22050-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This is a rabbit polyclonal antibody raised against the C terminus of human CEP97.
CEP97 (centrosomal protein of 97 kDa) has been identified as part of a complex with CP110. Together, CP110 and CEP97 have been shown to play a role in the assembly of cilia. Loss of CEP97 or CEP110 promotes cilia formation while forced expression inhibits ciliogenesis.Specifications
CEP97 | |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Canine | |
Q8IW35 | |
608540, 79598 | |
CEP97 Fusion Protein Ag16995 | |
Primary | |
-20°C | |
CEP97 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
2810403B08Rik, centrosomal protein 97kDa, Centrosomal protein of 97 kDa, CEP97, LRRIQ2 | |
CEP97 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CEP97 Rabbit anti-Canine, Human, Polyclonal, Proteintech