Learn More
Cesium hydroxide, 50% w/w aq. soln., 99% (metals basis)
Description
Cesium hydroxide is used in the preparation of porous cesium titanosilicates, which are potential ion-exchange materials utilized for radioactive wastes cleaning purpose. It is also involved in the preparation of other cesium salts. Further, it is used as an electrolyte in alkaline batteries and as a catalyst in organic synthesis. In addition to this, it is used in color photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 g |
UN Number | UN2681 |
Solubility Information | Miscible with ethanol. |
IUPAC Name | caesium(1+) hydroxide |
Formula Weight | 149.91 |
Percent Purity | 99% |
Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
Odor | Odorless |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
T26K015 | Certificate of Analysis | 13/08/2024 | 016505.MZ,016505.NF |
P09G016 | Certificate of Analysis | 23/12/2022 | 016505.MZ,016505.NF |
P25I011 | Certificate of Analysis | 12/04/2022 | 016505.MZ,016505.NF |
61500751 | Certificate of Analysis | 08/12/2021 | 016505.MZ,016505.NF |
T27H031 | Certificate of Analysis | 08/12/2021 | 016505.MZ,016505.NF |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.