missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFDP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CFDP1 |
---|---|
Concentration | 0.83 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885045
|
Proteintech
26636-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16875045
|
Proteintech
26636-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role during embryogenesis.Specifications
CFDP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UEE9 | |
10428 | |
CFDP1 Fusion Protein Ag24425 | |
Primary | |
-20°C | |
CFDP1 |
0.83 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BUCENTAUR, CFDP1, CP27, p97, SWC5, Yeti | |
CFDP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CFDP1 Rabbit anti-Human, Polyclonal, Proteintech