missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFHR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16583-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a secreted protein, which belongs to the complement factor H-related protein family. It binds to heparin, and may be involved in complement regulation. Mutations in this gene are associated with decreased risk of age-related macular degeneration, and with an increased risk of atypical hemolytic-uremic syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
CFHR3 | |
Polyclonal | |
Unconjugated | |
CFHR3 | |
CFHL3, CFHR3, complement factor H related 3, DOWN16, FHR 3, FHR3, H factor like protein 3, HLF4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10878 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02985 | |
CFHR3 | |
CFHR3 Fusion Protein Ag9963 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CFHR3 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction