missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Mouse, Clone: CFTR/1643, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1080-MSM3-P1
Additional Details : Weight : 0.01000kg
Specifications
CFTR | |
Cell Surface and Cytoplasmic | |
CFTR/1643 | |
Unconjugated | |
NP_011514056 | |
Mouse | |
165-170kDa | |
Protein A/G Purified | |
RUO | |
1080 | |
IgG2b |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CFTR | |
Recombinant human CFTR fragment (aa258-385) | |
MOLT-4 cells. Pancreas, Kidney or Placenta. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |