missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGGBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CGGBP1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897273
|
Proteintech
10716-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887273
|
Proteintech
10716-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CGGBP1 influences expression of the FMR1 gene which is associated with the fragile X mental retardation syndrome by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.Specifications
CGGBP1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q8BHG9, Q9UFW8 | |
106143, 288353, 8545 | |
CGGBP1 Fusion Protein Ag1129 | |
Primary | |
-20°C | |
CGGBP1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
20 kDa CGG binding protein, CGG binding protein 1, CGGBP, CGGBP1, p20 CGGBP, p20 CGGBP DNA binding protein | |
Cggbp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CGGBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech