Learn More
CHAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12963-1-AP-150UL
 View more versions of this product
Product Code. 16880944
  • 4652.90 SEK / 150µL
Description
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.
Specifications
Specifications
CHAD
Polyclonal
Unconjugated
CHAD
Cartilage leucine rich protein, CHAD, chondroadherin, SLRR4A
Rabbit
Antigen Affinity Chromatography
RUO
1101, 12643, 29195
-20°C
Liquid
Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15335, O55226, O70210
Chad
CHAD Fusion Protein Ag4077
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CHAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated