missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12963-1-AP-20UL
This item is not returnable.
View return policy
Description
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.Specifications
CHAD | |
Polyclonal | |
Unconjugated | |
CHAD | |
Cartilage leucine rich protein, CHAD, chondroadherin, SLRR4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1101, 12643, 29195 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15335, O55226, O70210 | |
Chad | |
CHAD Fusion Protein Ag4077 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHAD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction