missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD4 Mouse anti-Human, Rat, Clone: 3E4C6, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CHCHD4 |
---|---|
Clone | 3E4C6 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888855
|
Proteintech
66718-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878855
|
Proteintech
66718-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus.Specifications
CHCHD4 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CHCHD4, FLJ31709, MIA40 | |
Chchd4 | |
IgG1 | |
Protein G | |
Antibody |
3E4C6 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q8N4Q1 | |
131474, 312559 | |
CHCHD4 Fusion Protein Ag15339 | |
Primary | |
-20°C | |
CHCHD4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHCHD4 Mouse anti-Human, Rat, Clone: 3E4C6, Proteintech