Learn More
CHIT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21432-1-AP-150UL
 View more versions of this product
Product Code. 16881025
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.
Specifications
Specifications
CHIT1
Polyclonal
Unconjugated
CHIT1
CHI3, CHIT, CHIT1, Chitinase 1, chitinase 1 (chitotriosidase), Chitotriosidase 1
Rabbit
Antigen Affinity Chromatography
RUO
1118
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13231
CHIT1
CHIT1 Fusion Protein Ag16039
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHIT1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated