Learn More
CHIT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21432-1-AP-150UL
 View more versions of this product
Product Code. 16881025
  • 4652.90 SEK / 150µL
Description
Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.
Specifications
Specifications
CHIT1
Polyclonal
Unconjugated
CHIT1
CHI3, CHIT, CHIT1, Chitinase 1, chitinase 1 (chitotriosidase), Chitotriosidase 1
Rabbit
Antigen Affinity Chromatography
RUO
1118
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13231
CHIT1
CHIT1 Fusion Protein Ag16039
150 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CHIT1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated