missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25250-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.Specifications
CHL1 | |
Polyclonal | |
Unconjugated | |
CHL1 | |
CALL, CHL1, Close homolog of L1, L1CAM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10752, 12661 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00533, P70232 | |
CHL1 | |
CHL1 Fusion Protein Ag19263 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |