Learn More
CHL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25250-1-AP-150UL
 View more versions of this product
Product Code. 16873825
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known.
Specifications
Specifications
CHL1
Polyclonal
Unconjugated
CHL1
CALL, CHL1, Close homolog of L1, L1CAM2
Rabbit
Antigen Affinity Chromatography
RUO
10752, 12661
-20°C
Liquid
Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00533, P70232
CHL1
CHL1 Fusion Protein Ag19263
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated