Läs mer
Cholestane-3Beta,5Alpha,6Alpha-triol, TRC
Produktkod. 30366232
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366232 1 g
30349058 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366232 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4317451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35089-25-9
Molekylformel C27H48O3
Molekylvikt (g/mol) 420.67
Synonym 3beta-Hydroxycholestane-5alpha,6alpha-diol, (3beta,5alpha,6alpha)-Cholestane-3,5,6-triol
IUPAC-namn (3S,5R,6S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,3,4,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthrene-3,5,6-triol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3C[C@H](O)[C@@]4(O)C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Cholestane-3beta,5alpha,6alpha-triol
Kvantitet 1 g
InChI formel InChI=1S/C27H48O3/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24(29)27(30)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28-30H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24+,25-,26-,27+/m1/s1
Formel vikt 420.3603