missing translation for 'onlineSavingsMsg'
Learn More

CHOP/GADD153 Mouse anti-Human, Clone: 4F3G1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66741-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878895

  • 4721.67 SEK / 150µL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.
Specifications

Specifications

CHOP/GADD153
Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P35638
DDIT3
CHOP; GADD153 Fusion Protein Ag7354
150 μL
Primary
Human
Antibody
IgG2a
Western Blot
4F3G1
Unconjugated
DDIT3
C/EBP homologous protein, C/EBP homologous protein 10, CEBPZ, CHOP, CHOP 10, CHOP10, DDIT 3, DDIT3, GADD153
Mouse
Protein A
RUO
1649
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers