missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHOP/GADD153 Mouse anti-Human, Clone: 4F3G1, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CHOP/GADD153 |
---|---|
Clone | 4F3G1 |
Concentration | 1.5 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888895
|
Proteintech
66741-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878895
|
Proteintech
66741-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GADD153 is a small nuclear protein that is capable of dimerizing with transcription factors C/EBP alpha and beta. Once dimerized, this complex inhibits the normal binding and function of C/EBP to classical binding sites. Inversely, the C/EBP GADD153 dimer gains binding activity to other non classical C/EBP stress related targets. Under normal cellular conditions this protein is not expressed in detectable levels, but is highly unregulated during times of cellular/ER stress. Examples of GADD153 inducing stress include: treatment with tunicamycin, nutrient starvation and reducing agents that interfere with the calcium flux across the ER membrane.Specifications
CHOP/GADD153 | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C/EBP homologous protein, C/EBP homologous protein 10, CEBPZ, CHOP, CHOP 10, CHOP10, DDIT 3, DDIT3, GADD153 | |
DDIT3 | |
IgG2a | |
Protein A | |
Antibody |
4F3G1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P35638 | |
1649 | |
CHOP; GADD153 Fusion Protein Ag7354 | |
Primary | |
-20°C | |
DDIT3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CHOP/GADD153 Mouse anti-Human, Clone: 4F3G1, Proteintech