Learn More
Macherey-Nagel™ Chromabond™ Florisil Polypropylene SPE Column
Brand: Macherey-Nagel™ 730239
Additional Details : Weight : 1.00000kg
Specifications
6 mL | |
Solid Phase Extraction | |
Solid Phase Extraction Column |
2 g | |
Chromabond Florisil |