Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Particles M-270
RFP-Trap™ Magnetic Particles M-270 for immunoprecipitation (IP) of RFP-tagged proteins.
Brand:  Proteintech RTD-10
Additional Details : Weight : 0.00240kg
 View more versions of this product
Product Code. 17383443
  • 3400.00 SEK / 10 reactions
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
  • Consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet
Alpaca
IP, CoIP
Magnetic Particles M-270
10 Reactions