Learn More
Proteintech ChromoTek RFP-Trap™ Magnetic Particles M-270
RFP-Trap™ Magnetic Particles M-270 for immunoprecipitation (IP) of RFP-tagged proteins.
3400.00 SEK - 83240.00 SEK
Specifications
Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP
Target mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet
Label or Dye Magnetic Particles M-270
Species Alpaca
View More Specs
Products 5
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
17303453
View Documents Promotion Details
Proteintech
RTD-20
20 Reactions
5625.00 SEK
20 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
17313453
View Documents Promotion Details
Proteintech
RTD-200
200 Reactions
45670.00 SEK
200 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
17323453
View Documents Promotion Details
Proteintech
RTD-400
400 Reactions
83240.00 SEK
400 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
17383443
View Documents Promotion Details
Proteintech
RTD-10
10 Reactions
3400.00 SEK
10 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
17393443
View Documents Promotion Details
Proteintech
RTD-100
100 Reactions
25455.00 SEK
100 reactions
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • RFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP
  • Consists of a RFP VHH/ Nanobody coupled to Magnetic Particles M-270
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mRFP, mCherry, mRFPruby, mPlum, tagRFP, mKate2, mOrange, PA-mCherry, mScarlet
Alpaca
IP, CoIP
Magnetic Particles M-270
Videos
SDS
Dokumentation
Dokumentation
Zertifikate