Learn More
Proteintech ChromoTek SNAP/CLIP-tag-Trap™ Agarose Beads
SNAP/CLIP-tag™-Trap Agarose is an affinity reagent for IP of SNAP/CLIP-tag-fusion proteins.
Brand: Proteintech WTA-10
Additional Details : Weight : 0.00240kg
Description
- Consists of a SNAP/CLIP-tag Nanobody/ VHH, which is coupled to agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
SNAP-tag and CLIP-tag | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose | |
10 Reactions |