missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek SNAP/CLIP-tag-Trap™ Agarose Beads
SNAP/CLIP-tag™-Trap Agarose is an affinity reagent for IP of SNAP/CLIP-tag-fusion proteins.
2820.00 SEK - 69810.00 SEK
Specifications
Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | SNAP-tag and CLIP-tag |
Label or Dye | Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17303563
|
Proteintech
WTA-400 |
400 Reactions |
69810.00 SEK
400 reactions |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
17363553
|
Proteintech
WTA-10 |
10 Reactions |
2820.00 SEK
10 reactions |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
17373553
|
Proteintech
WTA-100 |
100 Reactions |
21280.00 SEK
100 reactions |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
17383553
|
Proteintech
WTA-20 |
20 Reactions |
4700.00 SEK
20 reactions |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
17393553
|
Proteintech
WTA-200 |
200 Reactions |
38310.00 SEK
200 reactions |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
- Consists of a SNAP/CLIP-tag Nanobody/ VHH, which is coupled to agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
SNAP-tag and CLIP-tag | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose |