Learn More
Proteintech ChromoTek SNAP/CLIP-tag-Trap™ Agarose Beads
SNAP/CLIP-tag™-Trap Agarose is an affinity reagent for IP of SNAP/CLIP-tag-fusion proteins.
Brand:  Proteintech WTA-200
Additional Details : Weight : 0.00240kg
 View more versions of this product
Product Code. 17393553
  • 38310.00 SEK / 200 reactions
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Consists of a SNAP/CLIP-tag Nanobody/ VHH, which is coupled to agarose beads
Specifications
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
SNAP-tag and CLIP-tag
Alpaca
IP, CoIP, ChIP, RIP
Agarose
200 Reactions