Läs mer
Cimetidine Sulfoxide, TRC
Produktkod. 30135016
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135016 1 g
30004780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135016 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4416801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54237-72-8
Molekylformel C10 H16 N6 O S
Molekylvikt (g/mol) 268.34
Synonym 2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine, Cimetidine Hydrochloride Imp. E (EP), Cimetidine Sulphoxide, Cimetidine Imp. E (EP)
IUPAC-namn 2-cyano-1-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfinyl]ethyl]guanidine
LEDER CNC(=NC#N)NCCS(=O)Cc1nc[nH]c1C
Specifikationer
Kemiskt namn eller material Cimetidine Sulfoxide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C10H16N6OS/c1-8-9(16-7-15-8)5-18(17)4-3-13-10(12-2)14-6-11/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14)
Formel vikt 268.1106
Procent renhet >95%