CAS RN 54237-72-8
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine (Cimetidine Sulphoxide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Cimetidine Sulfoxide, TRC
CAS: 54237-72-8 Molekylformel: C10 H16 N6 O S Molekylvikt (g/mol): 268.34 Synonym: 2-Cyano-1-methyl-3-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]sulphinyl]ethyl]guanidine,Cimetidine Hydrochloride Imp. E (EP),Cimetidine Sulphoxide,Cimetidine Imp. E (EP) IUPAC-namn: 2-cyano-1-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfinyl]ethyl]guanidine LEDER: CNC(=NC#N)NCCS(=O)Cc1nc[nH]c1C