Learn More
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 6.0
CAS: 6132-04-3294.10 g/mol
1090.00 SEK - 1670.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15486099
|
Thermo Scientific Chemicals
J63950.AK |
250 mL |
1090.00 SEK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15496099
|
Thermo Scientific Chemicals
J63950.AP |
500 mL |
1670.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6132-04-3 |
Specifications
Colorless | |
294.10 | |
6132-04-3 | |
Keep cold |
0.5M buffer soln. | |
6.0 | |
Odorless | |
Liquid |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 6.0